siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK134480.1 | - | 10 | 1292 | ||
| AK137466.1 | - | 10 | 1321 | |||
| AK153934.1 | - | 10 | 1265 | |||
| AK160874.1 | - | 10 | 1289 | |||
| AK167315.1 | - | 10 | 1264 | |||
| AK167375.1 | - | 10 | 1305 | |||
| AK210073.1 | - | 4 | 386 | |||
| BC007133.1 | - | 10 | 1284 | 3594358 | ||
| U35846.1 | - | 10 | 1249 | |||

Api5
Genomic Map