siRNA Details
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019915.2 | NP_064299.2 | 2 | 560 | ||
GenBank mRNA | AF016463.1 | - | 2 | 560 | ||
AF016465.1 | - | 2 | 560 | |||
X87612.1 | - | 2 | 560 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Art2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019915.2 | NP_064299.2 | 2 | 560 | ||
GenBank mRNA | AF016463.1 | - | 2 | 560 | ||
AF016465.1 | - | 2 | 560 | |||
X87612.1 | - | 2 | 560 |