siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080844.4 | NP_543120.1 | 5 | 857 | ||
| GenBank mRNA | AK050295.1 | - | 4 | 699 | ||
| BC019447.1 | - | 5 | 817 | 5125534 | ||
| BC033377.1 | - | 4 | 701 | 5123932 | ||
| S47225.1 | - | 4 | 660 | |||

Serpinc1
Genomic Map