siRNA Details
| Gene Symbol: Atp8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038999.2 | NP_001034088.1 | 28 | 2884 | ||
| NM_001284345.1 | NP_001271274.1 | 28 | 2884 | |||
| NM_009727.3 | NP_033857.1 | 27 | 2839 | |||
| GenBank mRNA | AK045367.1 | - | 28 | 2865 | ||
| AK076388.1 | - | 27 | 2775 | |||
| AK079430.1 | - | 6 | 682 | |||
| AK141559.1 | - | 28 | 2819 | |||
| AK220560.1 | - | 28 | 2794 | |||
| BC094235.1 | - | 17 | 1589 | |||
| BC129872.1 | - | 28 | 2659 | 40110214 | ||
| U75321.1 | - | 27 | 2618 | |||

Atp8a1
Genomic Map