siRNA Details
| Gene Symbol: Atp9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289445.1 | NP_001276374.1 | 26 | 3025 | ||
| NM_001289446.1 | NP_001276375.1 | 26 | 2755 | |||
| NM_015731.3 | NP_056546.2 | 26 | 2825 | |||
| GenBank mRNA | AF011336.1 | - | 25 | 2695 | ||
| AF152243.1 | - | 26 | 2993 | |||
| AK082271.1 | - | 26 | 2751 | |||
| AK089064.1 | - | 26 | 2931 | |||
| AK189628.1 | - | 1 | 26 | |||
| BC006949.1 | - | 24 | 2527 | |||
| BC075718.1 | - | 26 | 2953 | |||
| BC094551.1 | - | 26 | 2795 | 6813369 | ||

Atp9a
Genomic Map