siRNA Details
Gene Symbol: Atpif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF002718.1 | - | 3 | 264 | ||
AK008346.1 | - | 3 | 262 | |||
AK152389.1 | - | 4 | 1279 | |||
AV462016.1 | - | 3 | 325 | |||
BC012680.1 | - | 3 | 323 | 4217188 | ||
BU055998.1 | - | 3 | 251 | |||
CB848021.2 | - | 3 | 287 |
