siRNA Details
Gene Symbol: Avp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009732.1 | NP_033862.1 | 1 | 25 | ||
GenBank mRNA | AK138179.1 | - | 1 | 58 | ||
BC051997.1 | - | 1 | 38 | 5683720 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Avp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009732.1 | NP_033862.1 | 1 | 25 | ||
GenBank mRNA | AK138179.1 | - | 1 | 58 | ||
BC051997.1 | - | 1 | 38 | 5683720 |