siRNA Details
| Gene Symbol: Bcl7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009745.2 | NP_033875.2 | 4, 5 | 580 | ||
| GenBank mRNA | AJ011145.1 | - | 4, 5 | 460 | ||
| AK131643.1 | - | 4, 5 | 476 | |||
| AK155076.1 | - | 4, 5 | 462 | |||
| BC011471.1 | - | 4, 5 | 557 | 3660754 | ||

Bcl7b
Genomic Map