siRNA Details
| Gene Symbol: Bcl7c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009746.2 | NP_033876.1 | 4 | 543 | ||
| GenBank mRNA | AK131935.1 | - | 4 | 432 | ||
| BC005673.1 | - | 4 | 543 | 3153237 | ||
| BC058530.1 | - | 4 | 414 | 6818556 | ||
| Y11905.1 | - | 5 | 361 | |||

Bcl7c
Genomic Map