siRNA Details
| Gene Symbol: Bmp8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007559.4 | NP_031585.2 | 7 | 1353 | ||
| GenBank mRNA | AK082895.1 | - | 7 | 1354 | ||
| BC137890.1 | - | 7 | 1223 | 8860909 | ||
| U39545.1 | - | 7 | 1353 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp8b| Gene Symbol: Bmp8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007559.4 | NP_031585.2 | 7 | 1353 | ||
| GenBank mRNA | AK082895.1 | - | 7 | 1354 | ||
| BC137890.1 | - | 7 | 1223 | 8860909 | ||
| U39545.1 | - | 7 | 1353 | |||