siRNA Details
| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007562.2 | NP_031588.2 | 5 | 2478 | ||
| GenBank mRNA | AK087567.1 | - | 5 | 2473 | ||
| BC127226.1 | - | 1 | 145 | |||
| U88064.1 | - | 5 | 2425 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bnc1| Gene Symbol: Bnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007562.2 | NP_031588.2 | 5 | 2478 | ||
| GenBank mRNA | AK087567.1 | - | 5 | 2473 | ||
| BC127226.1 | - | 1 | 145 | |||
| U88064.1 | - | 5 | 2425 | |||