siRNA Details
| Gene Symbol: Bub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113179.1 | NP_001106650.1 | 8 | 821 | ||
| NM_009772.2 | NP_033902.2 | 8 | 818 | |||
| GenBank mRNA | AF002823.1 | - | 8 | 764 | ||
| AK021258.1 | - | 8 | 809 | |||
| AK042932.1 | - | 8 | 819 | |||
| AK144213.1 | - | 8 | 830 | |||
| BC029116.1 | - | 8 | 787 | 3671932 | ||
| U89795.1 | - | 8 | 836 | |||

Bub1
Genomic Map