siRNA Details
| Gene Symbol: C6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016704.2 | NP_057913.2 | 6, 7 | 852 | ||
| GenBank mRNA | AF184900.1 | - | 6, 7 | 853 | ||
| AK041057.1 | - | 6, 7 | 862 | |||
| BB222000.2 | - | 5, 6 | 604 | |||
| BC011251.1 | - | 6, 7 | 841 | 4196330 | ||

C6
Genomic Map