siRNA Details
| Gene Symbol: Cacna1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB086123.1 | - | 4 | 573 | ||
| AJ437291.1 | - | 4 | 555 | |||
| AJ437292.1 | - | 4 | 676 | |||
| AK147415.1 | - | 4 | 1051 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacna1d| Gene Symbol: Cacna1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB086123.1 | - | 4 | 573 | ||
| AJ437291.1 | - | 4 | 555 | |||
| AJ437292.1 | - | 4 | 676 | |||
| AK147415.1 | - | 4 | 1051 | |||