siRNA Details
| Gene Symbol: Cacybp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009786.2 | NP_033916.1 | 2 | 357 | ||
| GenBank mRNA | AK013924.1 | - | 2 | 288 | ||
| AK089043.1 | - | 2 | 211 | |||
| AK145013.1 | - | 2 | 324 | |||
| BC025948.1 | - | 2 | 242 | 5150537 | ||
| U97327.1 | - | 2 | 187 | |||

Cacybp
Genomic Map