siRNA Details
| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009791.4 | NP_033921.3 | 18 | 6562 | ||
| GenBank mRNA | AF533752.1 | - | 18 | 6540 | ||
| BC022903.1 | - | 1 | 882 | |||
| BC026699.1 | - | 2 | 2613 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aspm| Gene Symbol: Aspm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009791.4 | NP_033921.3 | 18 | 6562 | ||
| GenBank mRNA | AF533752.1 | - | 18 | 6540 | ||
| BC022903.1 | - | 1 | 882 | |||
| BC026699.1 | - | 2 | 2613 | |||