siRNA Details
| Gene Symbol: Capn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009794.3 | NP_033924.2 | 12 | 1514 | ||
| GenBank mRNA | AF015038.1 | - | 12 | 1460 | ||
| AK150695.1 | - | 12 | 1488 | |||
| AK152094.1 | - | 12 | 1488 | |||
| AK152790.1 | - | 12 | 1491 | |||
| AK152994.1 | - | 12 | 1491 | |||
| AK154855.1 | - | 12 | 1528 | |||
| BC054726.1 | - | 12 | 1514 | 5703563 | ||
| D38117.1 | - | 12 | 1555 | |||
| Y10139.1 | - | 12 | 1396 | |||

Capn2
Genomic Map