siRNA Details
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007619.2 | NP_031645.2 | 12 | 2126 | ||
GenBank mRNA | AK029826.1 | - | 12 | 2129 | ||
AK153915.1 | - | 12 | 2078 | |||
AK163907.1 | - | 11 | 1952 | |||
BC125285.1 | - | 12 | 2041 | 40129800 | ||
X57111.1 | - | 12 | 2013 |
