siRNA Details
| Gene Symbol: Cbl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007619.2 | NP_031645.2 | 13 | 2253 | ||
| GenBank mRNA | AK029826.1 | - | 13 | 2256 | ||
| AK153915.1 | - | 13 | 2205 | |||
| AK163907.1 | - | 12 | 2079 | |||
| BC125285.1 | - | 13 | 2168 | 40129800 | ||
| X57111.1 | - | 13 | 2140 | |||

Cbl
Genomic Map