siRNA Details
| Gene Symbol: Entpd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009849.2 | NP_033979.2 | 8 | 1311 | ||
| GenBank mRNA | AF042811.1 | - | 8 | 1235 | ||
| AK002553.1 | - | 8 | 1283 | |||
| AK172281.1 | - | 8 | 1259 | |||
| AY376711.1 | - | 8 | 1244 | |||
| BC011241.1 | - | 8 | 1243 | 3582550 | ||

Entpd2
Genomic Map