siRNA Details
| Gene Symbol: Cd247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113391.2 | NP_001106862.1 | 4 | 431 | ||
| NM_001113392.2 | NP_001106863.1 | 4 | 431 | |||
| NM_001113393.2 | NP_001106864.2 | 4 | 431 | |||
| NM_031162.4 | NP_112439.1 | 4 | 431 | |||
| GenBank mRNA | AK017904.1 | - | 4 | 394 | ||
| AK088087.1 | - | 4 | 359 | |||
| AK138774.1 | - | 4 | 432 | |||
| BC052824.1 | - | 4 | 352 | 30074812 | ||
| J04967.1 | - | 4 | 423 | |||
| L03353.1 | - | 4 | 347 | |||
| M33158.1 | - | 4 | 421 | |||
| S51932.1 | - | 4 | 347 | |||
| U56258.1 | - | 4 | 347 | |||
| X84237.1 | - | 4 | 347 | |||

Cd247
Genomic Map