siRNA Details
| Gene Symbol: Cd69 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033122.3 | NP_001028294.1 | 2 | 184 | ||
| GenBank mRNA | AK017979.1 | - | 2 | 185 | ||
| AK151076.1 | - | 2 | 185 | |||
| AK153224.1 | - | 2 | 185 | |||
| AK171645.1 | - | 1 | 3949 | |||
| BC106997.2 | - | 2 | 169 | |||
| BC128272.1 | - | 2 | 169 | 40081252 | ||
| BC128273.1 | - | 2 | 169 | 40081255 | ||
| BC139135.1 | - | 2 | 146 | 8862157 | ||
| BC139136.1 | - | 2 | 146 | 8862158 | ||

Cd69
Genomic Map