siRNA Details
| Gene Symbol: Cdk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009874.3 | NP_034004.2 | 10 | 921 | ||
| GenBank mRNA | AK145759.1 | - | 10 | 847 | ||
| AK168286.1 | - | 10 | 920 | |||
| BC004605.1 | - | 9 | 768 | 3585145 | ||
| BC068160.1 | - | 10 | 906 | 30358255 | ||
| BC141043.1 | - | 10 | 878 | 9055655 | ||
| U11822.1 | - | 10 | 884 | |||
| X74145.1 | - | 10 | 903 | |||

Cdk7
Genomic Map