siRNA Details
| Gene Symbol: Cdkn1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161624.1 | NP_001155096.1 | 2 | 412 | ||
| NM_009876.4 | NP_034006.3 | 2 | 374 | |||
| GenBank mRNA | AK077844.1 | - | 2 | 323 | ||
| AK132278.1 | - | 2 | 318 | |||
| BC005412.1 | - | 2 | 311 | 3490380 | ||
| CJ175313.1 | - | 2 | 374 | |||
| U20553.1 | - | 2 | 245 | |||
| U22399.1 | - | 2 | 303 | |||

Cdkn1c
Genomic Map