siRNA Details
| Gene Symbol: Cebpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024806.2 | NP_001019977.1 | 2 | 1410 | ||
| GenBank mRNA | AK136173.1 | - | 2 | 1391 | ||
| AK145374.1 | - | 2 | 1392 | |||
| AK155990.1 | - | 2 | 1393 | |||
| AK158771.1 | - | 2 | 1387 | |||
| AK159081.1 | - | 2 | 1407 | |||
| U19891.1 | - | 2 | 1381 | |||

Cebpz
Genomic Map