siRNA Details
Gene Symbol: Cfl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007688.2 | NP_031714.1 | 2 | 235 | ||
GenBank mRNA | AK004659.1 | - | 2 | 227 | ||
AK147174.1 | - | 2 | 149 | |||
BC007138.1 | - | 2 | 178 | 3590849 | ||
L29468.1 | - | 2 | 235 |
