siRNA Details
| Gene Symbol: Cflar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289704.2 | NP_001276633.1 | 9 | 1362 | ||
| NM_001293804.1 | NP_001280733.1 | 9 | 1345 | |||
| NM_001293805.1 | NP_001280734.1 | 7 | 838 | |||
| NM_207653.5 | NP_997536.1 | 9 | 1634 | |||
| GenBank mRNA | AK152515.1 | - | 9 | 1329 | ||
| AK161120.1 | - | 7 | 842 | |||
| BC029223.1 | - | 9 | 1603 | 4460554 | ||
| U97076.1 | - | 9 | 1300 | |||
| Y14041.1 | - | 8 | 1212 | |||

Cflar
Genomic Map