siRNA Details
| Gene Symbol: Chgb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007694.4 | NP_031720.1 | 4 | 2280 | ||
| GenBank mRNA | AK077487.1 | - | 4 | 1849 | ||
| AK077489.1 | - | 4 | 1888 | |||
| AK133629.1 | - | 4 | 1885 | |||
| AK133786.1 | - | 4 | 1890 | |||
| AK158857.1 | - | 4 | 2281 | |||
| BC014736.1 | - | 4 | 1881 | 4511248 | ||
| X51429.1 | - | 4 | 1876 | |||
| X53028.1 | - | 4 | 1886 | |||

Chgb
Genomic Map