siRNA Details
| Gene Symbol: Cish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009895.3 | NP_034025.1 | 3 | 674 | ||
| GenBank mRNA | AK077338.1 | - | 3 | 674 | ||
| AK086040.1 | - | 263 | ||||
| AK170116.1 | - | 2 | 249 | |||
| AK170410.1 | - | 2 | 252 | |||
| AK170554.1 | - | 3 | 475 | |||
| AK170877.1 | - | 3 | 470 | |||
| AK171431.1 | - | 3 | 475 | |||
| BC003783.1 | - | 3 | 478 | 3487580 | ||
| BC022178.1 | - | 3 | 439 | 4193243 | ||
| D31943.1 | - | 3 | 510 | |||

Cish
Genomic Map