siRNA Details
| Gene Symbol: Cish | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009895.3 | NP_034025.1 | 3 | 986 | ||
| GenBank mRNA | AK077338.1 | - | 3 | 986 | ||
| AK086040.1 | - | 575 | ||||
| AK170410.1 | - | 2 | 564 | |||
| AK170554.1 | - | 3 | 787 | |||
| AK171431.1 | - | 3 | 787 | |||

Cish
Genomic Map