siRNA Details
| Gene Symbol: Clcn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243762.1 | NP_001230691.1 | 4 | 656 | ||
| NM_016691.4 | NP_057900.3 | 2 | 274 | |||
| GenBank mRNA | AF134117.1 | - | 2 | 274 | ||
| AK052990.1 | - | 2 | 254 | |||
| AK142227.1 | - | 2 | 205 | |||
| AK145763.1 | - | 2 | 243 | |||
| BC036347.1 | - | 2 | 246 | 3989812 | ||

Clcn5
Genomic Map