siRNA Details
Gene Symbol: Cntn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159647.1 | NP_001153119.1 | 13 | 1777 | ||
NM_001159648.1 | NP_001153120.1 | 13 | 1631 | |||
NM_007727.2 | NP_031753.1 | 13 | 1680 | |||
GenBank mRNA | BC066864.1 | - | 13 | 1703 | 30099512 | |
X14943.1 | - | 13 | 1650 |
