siRNA Details
| Gene Symbol: Hps3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146323.1 | NP_001139795.1 | 9 | 1261 | ||
| NM_001146324.1 | NP_001139796.1 | 9 | 1657 | |||
| NM_080634.4 | NP_542365.3 | 9 | 1657 | |||
| GenBank mRNA | AF393747.1 | - | 9 | 1646 | ||
| AF393780.1 | - | 9 | 1649 | |||
| AK034377.1 | - | 9 | 1628 | |||
| AK036663.1 | - | 9 | 1665 | |||
| AK037829.1 | - | 8 | 1001 | |||
| AK049204.1 | - | 9 | 1650 | |||
| AK079896.1 | - | 9 | 1650 | |||
| AK086167.1 | - | 9 | 1621 | |||
| AK157953.1 | - | 9 | 1642 | |||
| AK169855.1 | - | 8 | 1729 | |||
| AK170581.1 | - | 9 | 1173 | |||
| AK171997.1 | - | 9 | 1249 | |||
| BC058235.1 | - | 9 | 1631 | 6307400 | ||

Hps3
Genomic Map