siRNA Details
| Gene Symbol: Col5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007737.2 | NP_031763.2 | 51 | 4047 | ||
| GenBank mRNA | AK084538.1 | - | 10 | 831 | ||
| AK084803.1 | - | 51 | 3685 | |||
| AK132413.1 | - | 51 | 3829 | |||
| AK139130.1 | - | 51 | 3622 | |||
| AK147220.1 | - | 51 | 3829 | |||
| AK147328.1 | - | 51 | 4048 | |||
| AK151929.1 | - | 51 | 3831 | |||
| AK160008.1 | - | 51 | 3621 | |||
| BC043696.1 | - | 25 | 1862 | |||
| BC055077.1 | - | 51 | 3798 | 30059991 | ||
| L02918.1 | - | 51 | 3679 | |||

Col5a2
Genomic Map