siRNA Details
| Gene Symbol: Col6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146007.2 | NP_666119.1 | 16 | 1584 | ||
| GenBank mRNA | AK044870.1 | - | 16 | 1506 | ||
| AK128938.1 | - | 16 | 1507 | |||
| AK132540.1 | - | 16 | 1506 | |||
| BC034414.1 | - | 16 | 1494 | 3594974 | ||
| L06343.1 | - | 12 | 607 | |||
| X62332.1 | - | 12 | 607 | |||
| X65582.1 | - | 15 | 1423 | |||

Col6a2
Genomic Map