siRNA Details
| Gene Symbol: Col1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007742.3 | NP_031768.2 | 5, 6 | 545 | ||
| GenBank mRNA | AK132180.1 | - | 5, 6 | 561 | ||
| AK158011.1 | - | 5, 6 | 562 | |||
| AK159283.1 | - | 5, 6 | 563 | |||
| AK159285.1 | - | 5, 6 | 565 | |||
| BC050014.1 | - | 5, 6 | 545 | 4225038 | ||
| BC059281.1 | - | 5, 6 | 555 | 3992363 | ||
| U08020.1 | - | 5, 6 | 446 | |||

Col1a1
Genomic Map