siRNA Details
| Gene Symbol: Copa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009938.4 | NP_034068.3 | 29 | 3434 | ||
| GenBank mRNA | AK032071.1 | - | 7 | 1387 | ||
| AK043479.1 | - | 29 | 3160 | |||
| AK181897.1 | - | 1 | 29 | |||
| BC005609.1 | - | 3 | 282 | |||
| BC024070.1 | - | 29 | 3087 | 5097228 | ||
| BC025896.1 | - | 11 | 1121 | |||
| BC047429.1 | - | 29 | 3125 | 3498656 | ||
| BC082785.1 | - | 25 | 2774 | |||

Copa
Genomic Map