siRNA Details
| Gene Symbol: Cox6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007748.3 | NP_031774.1 | 2 | 192 | ||
| GenBank mRNA | AK002402.1 | - | 2 | 194 | ||
| AK019318.1 | - | 2 | 197 | |||
| AK166496.1 | - | 2 | 194 | |||
| BC003807.1 | - | 2 | 192 | 3487598 | ||
| BC052816.1 | - | 2 | 198 | 6439987 | ||
| L06465.1 | - | 2 | 191 | |||
| U08440.1 | - | 2 | 181 | |||

Cox6a1
Genomic Map