siRNA Details
| Gene Symbol: Cryba4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021351.1 | NP_067326.1 | 4 | 383 | ||
| GenBank mRNA | AJ272228.2 | - | 4 | 383 | ||
| AK132591.1 | - | 5 | 400 | |||
| AK161601.1 | - | 3 | 347 | |||
| AK162018.1 | - | 5 | 397 | |||
| BC056444.1 | - | 4 | 373 | 6824277 | ||
| BC058703.1 | - | 4 | 383 | 6824293 | ||

Cryba4
Genomic Map