siRNA Details
Gene Symbol: Crygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144761.2 | NP_658906.1 | 3 | 342 | ||
GenBank mRNA | AK143110.1 | - | 3 | 341 | ||
BC056455.1 | - | 3 | 319 | 6824341 | ||
EF426303.1 | - | 3 | 327 | |||
K02585.1 | - | 2 | 197 |
