siRNA Details
| Gene Symbol: Crygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144761.2 | NP_658906.1 | 3 | 398 | ||
| GenBank mRNA | AK143110.1 | - | 3 | 397 | ||
| BC056455.1 | - | 3 | 375 | 6824341 | ||
| EF426303.1 | - | 3 | 383 | |||
| K02585.1 | - | 2 | 253 | |||

Crygb
Genomic Map