siRNA Details
| Gene Symbol: Crygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144761.2 | NP_658906.1 | 3 | 494 | ||
| GenBank mRNA | AK143110.1 | - | 3 | 493 | ||
| BC056455.1 | - | 3 | 471 | 6824341 | ||
| EF426303.1 | - | 3 | 479 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crygb| Gene Symbol: Crygb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144761.2 | NP_658906.1 | 3 | 494 | ||
| GenBank mRNA | AK143110.1 | - | 3 | 493 | ||
| BC056455.1 | - | 3 | 471 | 6824341 | ||
| EF426303.1 | - | 3 | 479 | |||