siRNA Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 11 | 1196 | ||
| GenBank mRNA | AK135114.1 | - | 4 | 1181 | ||
| BC150696.1 | - | 11 | 1136 | 9087607 | ||
| EF114317.1 | - | 10 | 1041 | |||
| EF114318.1 | - | 9 | 1275 | |||
| M85078.1 | - | 11 | 1137 | |||

Csf2ra
Genomic Map