siRNA Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 2 | 137 | ||
| GenBank mRNA | BC150696.1 | - | 2 | 77 | 9087607 | |
| EF114317.1 | - | 1 | 57 | |||
| EF114318.1 | - | 1 | 57 | |||
| M85078.1 | - | 2 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csf2ra| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 2 | 137 | ||
| GenBank mRNA | BC150696.1 | - | 2 | 77 | 9087607 | |
| EF114317.1 | - | 1 | 57 | |||
| EF114318.1 | - | 1 | 57 | |||
| M85078.1 | - | 2 | 78 | |||