siRNA Details
| Gene Symbol: Csf2ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009970.2 | NP_034100.2 | 4 | 383 | ||
| GenBank mRNA | BB664140.1 | - | 4 | 386 | ||
| BC150696.1 | - | 4 | 323 | 9087607 | ||
| BU614102.1 | - | 2 | 210 | |||
| EF114317.1 | - | 3 | 303 | |||
| EF114318.1 | - | 3 | 303 | |||
| M85078.1 | - | 4 | 324 | |||

Csf2ra
Genomic Map