siRNA Details
Gene Symbol: Csf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009971.1 | NP_034101.1 | 4, 5 | 526 | ||
GenBank mRNA | AK145177.1 | - | 4, 5 | 496 | ||
BC120759.1 | - | 4, 5 | 479 | 40129682 | ||
BC120761.1 | - | 4, 5 | 479 | 40129684 | ||
M13926.1 | - | 4, 5 | 526 |
