siRNA Details
Gene Symbol: Csn1s2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301334.1 | NP_001288263.1 | 2 | 70 | ||
NM_001301335.1 | NP_001288264.1 | 2 | 70 | |||
NM_009973.3 | NP_034103.2 | 2 | 70 | |||
GenBank mRNA | AK021337.1 | - | 2 | 66 | ||
AK021343.1 | - | 2 | 68 | |||
AK052780.1 | - | 2 | 68 | |||
AK052789.1 | - | 2 | 69 | |||
AK085744.1 | - | 2 | 70 | |||
AK085747.1 | - | 2 | 68 | |||
AK164763.1 | - | 2 | 68 | |||
AK164767.1 | - | 2 | 70 | |||
AK164768.1 | - | 2 | 68 | |||
AK164795.1 | - | 2 | 66 | |||
BC002084.1 | - | 2 | 51 | 3489886 |
