siRNA Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010016.2 | NP_034146.2 | 9, 10 | 1186 | ||
| GenBank mRNA | AF143541.1 | - | 11 | |||
| AK160994.1 | - | 9, 10 | 1164 | |||
| BC011314.1 | - | 9, 10 | 1141 | 3663561 | ||
| CT010328.1 | - | 9, 10 | 1115 | |||
| D63679.1 | - | 9, 10 | 1097 | |||
| L41366.1 | - | 9, 10 | 1130 | |||

Cd55
Genomic Map