siRNA Details
| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010016.2 | NP_034146.2 | 6 | 852 | ||
| GenBank mRNA | AK030285.1 | - | 6 | 844 | ||
| AK160994.1 | - | 6 | 830 | |||
| D63679.1 | - | 6 | 763 | |||
| L41366.1 | - | 6 | 796 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd55| Gene Symbol: Cd55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010016.2 | NP_034146.2 | 6 | 852 | ||
| GenBank mRNA | AK030285.1 | - | 6 | 844 | ||
| AK160994.1 | - | 6 | 830 | |||
| D63679.1 | - | 6 | 763 | |||
| L41366.1 | - | 6 | 796 | |||