siRNA Details
| Gene Symbol: Daf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007827.2 | NP_031853.2 | 10 | 1205 | ||
| GenBank mRNA | AK019553.1 | - | 2114 | |||
| BC115954.1 | - | 10 | 1205 | |||
| BC127150.1 | - | 8 | 876 | 40055987 | ||
| L41365.1 | - | 10 | 1214 | |||

Daf2
Genomic Map